siRNA Details
Gene Symbol: SYN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006950.3 | NP_008881.2 | 9 | 1213 | ||
NM_133499.2 | NP_598006.1 | 9 | 1213 | |||
GenBank mRNA | AL833961.1 | - | 1 | 36 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
