siRNA Details
Gene Symbol: FGF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004113.5 | NP_004104.3 | 6 | 1842 | ||
NM_021032.4 | NP_066360.1 | 5 | 2608 | |||
XM_005247227.2 | XP_005247284.1 | 5 | 1806 | |||
XM_006713538.3 | XP_006713601.1 | 5 | 1674 | |||
XM_017005879.1 | XP_016861368.1 | 5 | 1693 | |||
GenBank mRNA | BC022524.1 | - | 6 | 1826 | 4794600 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
