siRNA Details
Gene Symbol: GOLGA8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001023567.4 | NP_001018861.3 | 8 | 744 | ||
NR_027410.1 | 16 | 2992 | ||||
XM_011521596.1 | XP_011519898.1 | 15 | 1589 | |||
XM_011521597.1 | XP_011519899.1 | 14 | 1540 | |||
XM_017022210.1 | XP_016877699.1 | 16 | 1893 | |||
XM_017022211.1 | XP_016877700.1 | 16 | 1662 | |||
XM_017022212.1 | XP_016877701.1 | 15 | 2216 | |||
XM_017022213.1 | XP_016877702.1 | 15 | 1618 | |||
XM_017022214.1 | XP_016877703.1 | 16 | 1893 | |||
XM_017022215.1 | XP_016877704.1 | 16 | 2415 | |||
XM_017022216.1 | XP_016877705.1 | 15 | 3013 | |||
XM_017022217.1 | XP_016877706.1 | 14 | 2935 | |||
XM_017022218.1 | XP_016877707.1 | 16 | 3042 | |||
GenBank mRNA | BC143254.1 | - | 8 | 690 | 9051756 | |
BC144610.1 | - | 6 | 481 | 9053141 | ||
BX647673.1 | - | 15 | 2961 |
Gene Symbol: GOLGA8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181077.3 | NP_851422.1 | 8 | 744 | ||
NR_027409.1 | 16 | 3006 | ||||
XM_006720440.2 | XP_006720503.1 | 8 | 712 | |||
XM_017022018.1 | XP_016877507.1 | 12 | 3140 | |||
XM_017022019.1 | XP_016877508.1 | 11 | 2836 | |||
XM_017022020.1 | XP_016877509.1 | 6 | 834 | |||
GenBank mRNA | AB020662.1 | - | 7 | 724 | ||
AF163441.1 | - | 6 | 1168 | |||
AF204231.1 | - | 7 | 1061 | |||
AL137686.1 | - | 6 | 468 | |||
BC032774.1 | - | 8 | 646 | |||
BC093673.1 | - | 8 | 690 | 7939518 | ||
BC104800.1 | - | 8 | 690 | 8143803 | ||
BC144609.1 | - | 6 | 481 | 9053140 | ||
BC150329.1 | - | 7 | 735 | |||
BC152410.1 | - | 7 | 724 | |||
BX648160.1 | - | 15 | 2960 | |||
BX648575.1 | - | 15 | 2713 | |||
CR749361.1 | - | 8 | 1897 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
