siRNA Details
Gene Symbol: CTDSPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008392.1 | NP_001008393.1 | 8 | 1281 | ||
NM_005808.2 | NP_005799.2 | 7 | 1248 | |||
GenBank mRNA | D88153.1 | - | 8 | 1595 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
