siRNA Details
Gene Symbol: GYPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304382.1 | NP_001291311.1 | 4 | 289 | ||
NM_002100.5 | NP_002091.3 | 3 | 209 | |||
XM_011531903.2 | XP_011530205.1 | 3 | 344 | |||
XM_011531904.2 | XP_011530206.1 | 4 | 2848 | |||
XM_011531905.2 | XP_011530207.1 | 4 | 496 | |||
XM_017008137.1 | XP_016863626.1 | 3 | 2543 | |||
GenBank mRNA | AY950609.1 | - | 3 | 151 | ||
AY950611.1 | - | 3 | 151 | |||
BC069310.1 | - | 3 | 201 | 7262104 | ||
BC121077.1 | - | 3 | 185 | 40119713 | ||
BC121078.1 | - | 3 | 185 | 40119716 | ||
EU338223.1 | - | 2 | 89 | |||
EU338224.1 | - | 4 | 312 | |||
EU338225.1 | - | 4 | 281 | |||
EU338226.1 | - | 4 | 246 | |||
EU338227.1 | - | 2 | 89 | |||
EU338228.1 | - | 2 | 89 | |||
EU338229.1 | - | 4 | 312 | |||
EU338230.1 | - | 4 | 312 | |||
EU338235.1 | - | 2 | 89 | |||
EU338236.1 | - | 4 | 312 | |||
EU338237.1 | - | 5 | 405 | |||
J02982.1 | - | 3 | 161 | |||
X08055.1 | - | 5 | 287 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
