siRNA Details
Gene Symbol: EYA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282560.1 | NP_001269489.1 | 2, 3 | 215 | ||
NM_001282561.1 | NP_001269490.1 | 2, 3 | 215 | |||
NM_001282562.1 | NP_001269491.1 | 2, 3 | 215 | |||
NM_001990.3 | NP_001981.2 | 2, 3 | 215 | |||
NR_104214.1 | 2, 3 | 215 | ||||
XM_011541001.2 | XP_011539303.1 | 2, 3 | 212 | |||
XM_017000654.1 | XP_016856143.1 | 2, 3 | 212 | |||
XM_017000655.1 | XP_016856144.1 | 3, 4 | 368 | |||
XM_017000657.1 | XP_016856146.1 | 2, 3 | 245 | |||
XM_017000658.1 | XP_016856147.1 | 2, 3 | 212 | |||
XM_017000659.1 | XP_016856148.1 | 2, 3 | 209 | |||
GenBank mRNA | AK289805.1 | - | 2, 3 | 174 | ||
AK295745.1 | - | 2, 3 | 209 | |||
AK298129.1 | - | 2, 3 | 212 | |||
AK303664.1 | - | 2, 3 | 209 | |||
AL603098.1 | - | 2, 3 | 198 | |||
BC029500.1 | - | 2, 3 | 175 | 5272319 | ||
BC041667.1 | - | 2, 3 | 142 | 5743906 | ||
BX648945.1 | - | 2, 3 | 184 | |||
DC416048.1 | - | 2, 3 | 215 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
