siRNA Details
Gene Symbol: EYA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282560.1 | NP_001269489.1 | 14 | 1390 | ||
NM_001282561.1 | NP_001269490.1 | 14 | 1390 | |||
NM_001282562.1 | NP_001269491.1 | 14 | 1448 | |||
NM_001990.3 | NP_001981.2 | 15 | 1528 | |||
NR_104214.1 | 16 | 1596 | ||||
XM_006710449.3 | XP_006710512.1 | 16 | 1564 | |||
XM_011540999.2 | XP_011539301.1 | 16 | 1567 | |||
XM_011541001.2 | XP_011539303.1 | 15 | 1525 | |||
XM_011541003.2 | XP_011539305.1 | 15 | 1427 | |||
XM_011541004.2 | XP_011539306.1 | 15 | 1424 | |||
XM_017000652.1 | XP_016856141.1 | 16 | 1568 | |||
XM_017000653.1 | XP_016856142.1 | 17 | 1688 | |||
XM_017000654.1 | XP_016856143.1 | 15 | 1525 | |||
XM_017000655.1 | XP_016856144.1 | 16 | 1681 | |||
XM_017000656.1 | XP_016856145.1 | 15 | 1428 | |||
XM_017000657.1 | XP_016856146.1 | 14 | 1420 | |||
XM_017000658.1 | XP_016856147.1 | 14 | 1445 | |||
XM_017000659.1 | XP_016856148.1 | 13 | 1304 | |||
GenBank mRNA | AJ007991.1 | - | 2 | 110 | ||
AK289805.1 | - | 15 | 1487 | |||
AK295745.1 | - | 14 | 1384 | |||
AK298129.1 | - | 14 | 1445 | |||
AK303664.1 | - | 14 | 1372 | |||
BC029500.1 | - | 16 | 1556 | 5272319 | ||
BC041667.1 | - | 14 | 1317 | 5743906 | ||
BX648945.1 | - | 15 | 1497 | |||
U81602.1 | - | 10 | 1127 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
