siRNA Details
Gene Symbol: CLEC2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004419.4 | NP_001004419.1 | 3 | 237 | ||
NM_001197317.2 | NP_001184246.1 | 2 | 126 | |||
NM_001197318.2 | NP_001184247.1 | 3 | 237 | |||
NM_001197319.2 | NP_001184248.1 | 2 | 126 | |||
NM_013269.5 | NP_037401.1 | 3 | 237 | |||
NR_036693.2 | 3 | 237 | ||||
GenBank mRNA | AF133299.1 | - | 3 | 223 | ||
AF285087.1 | - | 3 | 215 | |||
AF285088.1 | - | 2 | 132 | |||
AF285089.1 | - | 2 | 147 | |||
AK310150.1 | - | 3 | 233 | |||
AK310554.1 | - | 3 | 228 | |||
AK313554.1 | - | 3 | 228 | |||
BC019883.1 | - | 3 | 217 | 4524891 | ||
BU929933.1 | - | 3 | 222 | |||
DA814760.1 | - | 3 | 232 | |||
DB111977.1 | - | 3 | 238 | |||
DB124897.1 | - | 3 | 233 | |||
FN813349.1 | - | 3 | 210 | |||
FN813350.1 | - | 2 | 99 | |||
FN813351.1 | - | 2 | 99 | |||
KF958454.1 | - | 3 | 210 | |||
KF958455.1 | - | 3 | 210 | |||
KF958456.1 | - | 3 | 210 | |||
KF958457.1 | - | 3 | 210 | |||
KF958458.1 | - | 3 | 210 | |||
KF958459.1 | - | 2 | 99 | |||
KF971856.1 | - | 3 | 210 | |||
KF971857.1 | - | 3 | 210 | |||
KF971858.1 | - | 2 | 99 | |||
KF971859.1 | - | 3 | 210 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
