siRNA Details
Gene Symbol: HBD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000519.3 | NP_000510.1 | 2 | 362 | ||
GenBank mRNA | AY034468.1 | - | 2 | 362 | ||
BC069307.1 | - | 2 | 217 | 7262103 | ||
BC070282.1 | - | 2 | 218 | 30418964 | ||
BU664913.1 | - | 2 | 181 |
Gene Symbol: HBB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000518.4 | NP_000509.1 | 2 | 217 | ||
GenBank mRNA | AF117710.1 | - | 2 | 197 | ||
AF181832.1 | - | 2 | 110 | |||
AF181989.1 | - | 2 | 200 | |||
AF349114.1 | - | 2 | 202 | |||
AK311825.1 | - | 2 | 217 | |||
AY136510.1 | - | 2 | 167 | |||
AY509193.1 | - | 2 | 217 | |||
BC007075.1 | - | 2 | 218 | 4292125 | ||
CR536530.1 | - | 2 | 167 | |||
CR541913.1 | - | 2 | 167 | |||
DQ655983.2 | - | 2 | 98 | |||
EU694432.1 | - | 2 | 214 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
