siRNA Details
Gene Symbol: C17orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142798.2 | NP_001136270.1 | 4 | 365 | ||
NM_001142799.2 | NP_001136271.1 | 3 | 263 | |||
NM_174893.3 | NP_777553.1 | 4 | 365 | |||
GenBank mRNA | AK055800.1 | - | 2 | 1584 | ||
AK295795.1 | - | 4 | 320 | |||
BC040036.1 | - | 4 | 297 | 5406966 | ||
BM011108.1 | - | 4 | 328 | |||
BU596784.1 | - | 4 | 368 | |||
CB119197.1 | - | 4 | 348 | |||
CN280697.1 | - | 3 | 231 |
Gene Symbol: RNASEK-C17orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037717.1 | 6 | 1375 | |||
GenBank mRNA | BC040148.1 | - | 6 | 1132 | 5588971 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
