siRNA Details
Gene Symbol: JMJD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005920.2 | NP_001005920.2 | 7 | 683 | ||
NM_001323918.1 | NP_001310847.1 | 7 | 690 | |||
NM_001323919.1 | NP_001310848.1 | 7 | 690 | |||
NM_001323920.1 | NP_001310849.1 | 6 | 600 | |||
NM_001323922.1 | NP_001310851.1 | 6 | 600 | |||
NR_136650.1 | 6 | 778 | ||||
NR_136651.1 | 6 | 778 | ||||
NR_136652.1 | 5 | 688 | ||||
GenBank mRNA | AF370420.1 | - | 7 | 567 | ||
AL524336.3 | - | 6 | 663 | |||
AL524922.3 | - | 6 | 665 | |||
BC073785.1 | - | 6 | 655 | |||
BC094850.1 | - | 4 | 301 | |||
BC137100.1 | - | 7 | 649 | 9021097 | ||
BC137101.1 | - | 7 | 649 | 9021098 | ||
BG284477.1 | - | 2 | 140 | |||
BG423721.1 | - | 3 | 200 | |||
BG481039.1 | - | 7 | 586 | |||
BG826580.1 | - | 4 | 481 | |||
BI463867.1 | - | 7 | 690 | |||
BI767435.1 | - | 6 | 503 | |||
BX335015.2 | - | 7 | 569 | |||
DQ074695.1 | - | 6 | 473 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
