siRNA Details
Gene Symbol: SDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014654.3 | NP_055469.3 | 5 | 1360 | ||
XM_011542463.1 | XP_011540765.1 | 5 | 1295 | |||
XM_011542464.2 | XP_011540766.1 | 5 | 1565 | |||
XM_011542466.1 | XP_011540768.1 | 5 | 1202 | |||
GenBank mRNA | AB007937.1 | - | 3 | 2642 | ||
AF248634.1 | - | 5 | 1367 | |||
AK295698.1 | - | 4 | 1105 | |||
BQ887307.1 | - | 3 | 678 | |||
KU178484.1 | - | 4 | 1097 | |||
KU178485.1 | - | 3 | 1139 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
