siRNA Details
Gene Symbol: RPL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000661.4 | NP_000652.2 | 8 | 670 | ||
NM_001024921.3 | NP_001020092.1 | 8 | 828 | |||
GenBank mRNA | AB062431.1 | - | 1 | 617 | ||
AK130966.1 | - | 1 | 633 | |||
AK303908.1 | - | 2 | 574 | |||
AL832047.1 | - | 3 | 4226 | |||
AY320412.1 | - | 8 | 653 | |||
AY376242.1 | - | 7 | 787 | |||
AY597415.1 | - | 1 | 644 | |||
BC000483.2 | - | 1 | 622 | 2964733 | ||
BC004156.2 | - | 1 | 616 | 2961026 | ||
BC004206.2 | - | 1 | 616 | 2961026 | ||
BC007967.2 | - | 1 | 672 | 4304670 | ||
BC012149.1 | - | 1 | 672 | 4554838 | ||
BC031906.1 | - | 7 | 617 | 4996986 | ||
BC066318.1 | - | 1 | 637 | 5277491 | ||
BC070214.1 | - | 1 | 676 | 6605055 | ||
BC100285.1 | - | 7 | 777 | 6022858 | ||
BG829769.1 | - | 7 | 617 | |||
D14531.1 | - | 7 | 617 | |||
U09953.1 | - | 8 | 640 | |||
U21138.1 | - | 1 | 617 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
