siRNA Details
Gene Symbol: SMARCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289396.1 | NP_001276325.1 | 34 | 5296 | ||
NM_001289397.1 | NP_001276326.1 | 33 | 5081 | |||
NM_001289398.1 | NP_001276327.1 | 7 | 1263 | |||
NM_001289399.1 | NP_001276328.1 | 8 | 1250 | |||
NM_001289400.1 | NP_001276329.1 | 8 | 1671 | |||
NM_003070.4 | NP_003061.3 | 34 | 5309 | |||
NM_139045.3 | NP_620614.2 | 33 | 5255 | |||
GenBank mRNA | AK094076.1 | - | 8 | 1671 | ||
AK298045.1 | - | 8 | 1250 | |||
X72889.1 | - | 34 | 5297 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
