siRNA Details
Gene Symbol: SH3D19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009555.3 | NP_001009555.3 | 15 | 2714 | ||
NM_001128923.1 | NP_001122395.1 | 14 | 2633 | |||
NM_001128924.1 | NP_001122396.1 | 7 | 1636 | |||
NM_001243349.1 | NP_001230278.1 | 14 | 2554 | |||
XM_005262767.2 | XP_005262824.1 | 16 | 2575 | |||
XM_011531646.1 | XP_011529948.1 | 14 | 2361 | |||
XM_011531647.1 | XP_011529949.1 | 13 | 2253 | |||
XM_011531648.1 | XP_011529950.1 | 16 | 2688 | |||
XM_011531649.1 | XP_011529951.1 | 12 | 2184 | |||
XM_017007778.1 | XP_016863267.1 | 16 | 2679 | |||
XM_017007779.1 | XP_016863268.1 | 12 | 2175 | |||
XM_017007780.1 | XP_016863269.1 | 15 | 2610 | |||
XM_017007781.1 | XP_016863270.1 | 15 | 2605 | |||
XM_017007782.1 | XP_016863271.1 | 14 | 2536 | |||
XM_017007783.1 | XP_016863272.1 | 15 | 2506 | |||
GenBank mRNA | AK092528.1 | - | 7 | 866 | ||
AK128493.1 | - | 7 | 1485 | |||
AK291025.1 | - | 7 | 802 | |||
AK294476.1 | - | 14 | 2554 | |||
AL133047.1 | - | 5 | 478 | |||
BC010110.2 | - | 7 | 907 | |||
BC085613.1 | - | 6 | 693 | |||
BC108890.1 | - | 8 | 1744 | 40001764 | ||
BC108891.1 | - | 8 | 1744 | 40001765 | ||
BC108892.1 | - | 8 | 1744 | 40001766 | ||
BC108893.1 | - | 7 | 1636 | 40001767 | ||
BX647422.1 | - | 15 | 2626 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
