siRNA Details
Gene Symbol: SH3D19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009555.3 | NP_001009555.3 | 21 | 4945 | ||
NM_001128923.1 | NP_001122395.1 | 20 | 4864 | |||
NM_001128924.1 | NP_001122396.1 | 13 | 3867 | |||
NM_001243349.1 | NP_001230278.1 | 20 | 4785 | |||
XM_005262767.2 | XP_005262824.1 | 22 | 4806 | |||
XM_011531646.1 | XP_011529948.1 | 20 | 4592 | |||
XM_011531647.1 | XP_011529949.1 | 19 | 4484 | |||
XM_011531648.1 | XP_011529950.1 | 22 | 4919 | |||
XM_011531649.1 | XP_011529951.1 | 18 | 4415 | |||
XM_017007778.1 | XP_016863267.1 | 22 | 4910 | |||
XM_017007779.1 | XP_016863268.1 | 18 | 4406 | |||
XM_017007780.1 | XP_016863269.1 | 21 | 4841 | |||
XM_017007781.1 | XP_016863270.1 | 21 | 4836 | |||
XM_017007782.1 | XP_016863271.1 | 20 | 4767 | |||
XM_017007783.1 | XP_016863272.1 | 21 | 4737 | |||
GenBank mRNA | AK128493.1 | - | 13 | 3712 | ||
AK294476.1 | - | 20 | 4781 | |||
AL133047.1 | - | 11 | 2706 | |||
BC032468.1 | - | 3 | 1794 | |||
BC045742.1 | - | 5 | 2106 | |||
BC085613.1 | - | 12 | 2920 | |||
BM831166.1 | - | 1 | 477 | |||
BX647422.1 | - | 21 | 4853 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
