siRNA Details
Gene Symbol: COL4A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000091.4 | NP_000082.2 | 35 | 3100 | ||
XM_005246277.3 | XP_005246334.1 | 34 | 2971 | |||
XM_005246280.3 | XP_005246337.1 | 35 | 3076 | |||
XM_006712245.3 | XP_006712308.1 | 35 | 3076 | |||
XM_011510555.1 | XP_011508857.1 | 35 | 3076 | |||
XM_011510556.2 | XP_011508858.1 | 22 | 2132 | |||
XM_017003295.1 | XP_016858784.1 | 35 | 3076 | |||
GenBank mRNA | BM712932.1 | - | 2 | 127 | ||
X80031.1 | - | 35 | 3099 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
