siRNA Details
Gene Symbol: NUP58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014089.3 | NP_054808.1 | 3 | 513 | ||
XM_005266600.3 | XP_005266657.1 | 3 | 857 | |||
XM_011535328.1 | XP_011533630.1 | 3 | 857 | |||
XM_011535329.2 | XP_011533631.1 | 4 | 943 | |||
XM_017020880.1 | XP_016876369.1 | 5 | 1025 | |||
XM_017020881.1 | XP_016876370.1 | 4 | 943 | |||
GenBank mRNA | AB007870.1 | - | 3 | 410 | ||
AK123955.1 | - | 4 | 347 | |||
BC001104.2 | - | 3 | 385 | 3504343 | ||
BP249090.1 | - | 3 | 513 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
