siRNA Details
Gene Symbol: NUP58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008564.1 | NP_001008564.1 | 9 | 1187 | ||
NM_014089.3 | NP_054808.1 | 10 | 1223 | |||
XM_005266600.3 | XP_005266657.1 | 10 | 1567 | |||
XM_011535328.1 | XP_011533630.1 | 10 | 1567 | |||
XM_011535329.2 | XP_011533631.1 | 11 | 1653 | |||
XM_017020880.1 | XP_016876369.1 | 12 | 1735 | |||
XM_017020881.1 | XP_016876370.1 | 10 | 1628 | |||
GenBank mRNA | AB007870.1 | - | 10 | 1120 | ||
AK123955.1 | - | 11 | 1057 | |||
AK302946.1 | - | 9 | 1082 | |||
AK303409.1 | - | 9 | 991 | |||
BC001104.2 | - | 10 | 1095 | 3504343 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
