siRNA Details
Gene Symbol: RALGAPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283043.1 | NP_001269972.1 | 42 | 7159 | ||
NM_014990.1 | NP_055805.1 | 41 | 7120 | |||
NM_194301.2 | NP_919277.2 | 40 | 7089 | |||
XM_005267491.3 | XP_005267548.1 | 42 | 7311 | |||
XM_005267493.3 | XP_005267550.1 | 41 | 7170 | |||
XM_005267498.4 | XP_005267555.1 | 27 | 4921 | |||
XM_006720098.2 | XP_006720161.1 | 42 | 8688 | |||
XM_006720099.2 | XP_006720162.1 | 41 | 8685 | |||
XM_006720100.3 | XP_006720163.1 | 41 | 8547 | |||
XM_006720101.2 | XP_006720164.1 | 43 | 8719 | |||
XM_006720102.3 | XP_006720165.1 | 42 | 8422 | |||
XM_006720103.2 | XP_006720166.1 | 43 | 7342 | |||
XM_017021141.1 | XP_016876630.1 | 42 | 8578 | |||
XM_017021142.1 | XP_016876631.1 | 41 | 8575 | |||
XM_017021144.1 | XP_016876633.1 | 42 | 7339 | |||
XM_017021145.1 | XP_016876634.1 | 42 | 7201 | |||
XM_017021146.1 | XP_016876635.1 | 27 | 6357 | |||
XM_017021147.1 | XP_016876636.1 | 28 | 5274 | |||
GenBank mRNA | AK126975.1 | - | 12 | 2621 | ||
AL834362.1 | - | 18 | 3497 | |||
AY596970.1 | - | 41 | 7120 | |||
AY596971.1 | - | 40 | 7089 | |||
BC042013.1 | - | 10 | 1614 | |||
BC042045.1 | - | 18 | 3291 |
Gene Symbol: RALGAPA1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104269.1 | 1 | 7153 | |||
GenBank mRNA | AL050050.1 | - | 1 | 1765 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
