siRNA Details
Gene Symbol: RALGAPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283043.1 | NP_001269972.1 | 42 | 7741 | ||
NM_014990.1 | NP_055805.1 | 41 | 7702 | |||
NM_194301.2 | NP_919277.2 | 40 | 7671 | |||
XM_006720098.2 | XP_006720161.1 | 42 | 9270 | |||
XM_006720099.2 | XP_006720162.1 | 41 | 9267 | |||
XM_006720101.2 | XP_006720164.1 | 43 | 9301 | |||
XM_006720103.2 | XP_006720166.1 | 43 | 7924 | |||
XM_017021141.1 | XP_016876630.1 | 42 | 9160 | |||
XM_017021145.1 | XP_016876634.1 | 42 | 7783 | |||
XM_017021147.1 | XP_016876636.1 | 28 | 5856 | |||
GenBank mRNA | AK126975.1 | - | 12 | 3203 | ||
AL834362.1 | - | 18 | 4079 | |||
AY596970.1 | - | 41 | 7702 | |||
AY596971.1 | - | 40 | 7671 | |||
BC042013.1 | - | 10 | 2196 | |||
DQ786317.1 | - | 5 | 1772 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
