siRNA Details
Gene Symbol: CGGBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008390.1 | NP_001008391.1 | 4 | 945 | ||
NM_001195308.1 | NP_001182237.1 | 4 | 926 | |||
NM_003663.3 | NP_003654.3 | 3 | 843 | |||
XM_017007355.1 | XP_016862844.1 | 3 | 751 | |||
GenBank mRNA | AJ000258.1 | - | 3 | 629 | ||
AL117392.1 | - | 3 | 789 | |||
BC052980.1 | - | 3 | 745 | 6012191 | ||
CR456854.1 | - | 1 | 433 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
