siRNA Details
Gene Symbol: MLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015166.3 | NP_055981.1 | 12 | 3305 | ||
NM_139202.2 | NP_631941.1 | 12 | 2944 | |||
XM_011530678.1 | XP_011528980.1 | 11 | 2901 | |||
XM_017028671.1 | XP_016884160.1 | 11 | 2733 | |||
GenBank mRNA | AF319633.1 | - | 12 | 2867 | ||
AK124264.1 | - | 14 | 2925 | |||
BC010518.1 | - | 1 | 466 | |||
BC028425.2 | - | 12 | 2904 | 4817175 | ||
BC070042.1 | - | 12 | 2902 | 5285102 | ||
D25217.3 | - | 12 | 2981 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
