siRNA Details
Gene Symbol: EPSTI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002264.2 | NP_001002264.1 | 2 | 313 | ||
NM_033255.3 | NP_150280.1 | 2 | 313 | |||
XM_005266596.1 | XP_005266653.1 | 2 | 313 | |||
XM_005266597.1 | XP_005266654.1 | 2 | 313 | |||
XM_006719896.3 | XP_006719959.1 | 2 | 313 | |||
XM_011535312.1 | XP_011533614.1 | 2 | 1523 | |||
XM_017020853.1 | XP_016876342.1 | 2 | 103 | |||
XM_017020854.1 | XP_016876343.1 | 3 | 689 | |||
GenBank mRNA | AF396928.1 | - | 2 | 272 | ||
AK292461.1 | - | 2 | 283 | |||
AK313877.1 | - | 2 | 277 | |||
AL831953.1 | - | 2 | 271 | |||
BC023660.2 | - | 2 | 272 | 4865634 | ||
BP228419.1 | - | 3 | 282 | |||
BU603558.1 | - | 286 | ||||
DA940572.1 | - | 2 | 314 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
