siRNA Details
Gene Symbol: SH2D3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142531.1 | NP_001136003.1 | 3 | 115 | ||
NM_001142532.1 | NP_001136004.1 | 3 | 364 | |||
NM_001142533.1 | NP_001136005.1 | 2 | 252 | |||
NM_001142534.1 | NP_001136006.1 | 2 | 224 | |||
NM_001252334.1 | NP_001239263.1 | 4 | 621 | |||
NM_005489.3 | NP_005480.2 | 3 | 250 | |||
NM_170600.2 | NP_733745.1 | 4 | 723 | |||
XM_005251639.1 | XP_005251696.1 | 3 | 231 | |||
XM_011518114.1 | XP_011516416.1 | 4 | 707 | |||
XM_011518115.1 | XP_011516417.1 | 4 | 667 | |||
XM_011518117.2 | XP_011516419.1 | 2 | 1343 | |||
XM_017014174.1 | XP_016869663.1 | 2 | 504 | |||
GenBank mRNA | AF124251.1 | - | 3 | 162 | ||
AK056068.1 | - | 2 | 252 | |||
AK128287.1 | - | 5 | 654 | |||
AK291393.1 | - | 2 | 216 | |||
AK292041.1 | - | 3 | 178 | |||
AK292718.1 | - | 4 | 594 | |||
AK294386.1 | - | 3 | 115 | |||
AK311510.1 | - | 3 | 364 | |||
AY358089.1 | - | 3 | 232 | |||
AY358440.1 | - | 3 | 151 | |||
BC027962.1 | - | 3 | 121 | 5223012 | ||
BC032365.1 | - | 4 | 598 | 4521962 | ||
BI912337.1 | - | 3 | 319 | |||
BX647905.1 | - | 4 | 547 | |||
DA970566.1 | - | 3 | 223 | |||
DC302682.1 | - | 2 | 224 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
