siRNA Details
Gene Symbol: SH2D3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142531.1 | NP_001136003.1 | 7 | 1374 | ||
NM_001142532.1 | NP_001136004.1 | 7 | 1623 | |||
NM_001142533.1 | NP_001136005.1 | 6 | 1511 | |||
NM_001142534.1 | NP_001136006.1 | 6 | 1483 | |||
NM_001252334.1 | NP_001239263.1 | 8 | 1880 | |||
NM_005489.3 | NP_005480.2 | 7 | 1509 | |||
NM_170600.2 | NP_733745.1 | 8 | 1982 | |||
XM_005251639.1 | XP_005251696.1 | 7 | 1490 | |||
XM_011518114.1 | XP_011516416.1 | 8 | 1966 | |||
XM_011518115.1 | XP_011516417.1 | 8 | 1926 | |||
XM_011518117.2 | XP_011516419.1 | 6 | 2602 | |||
XM_017014174.1 | XP_016869663.1 | 6 | 1763 | |||
GenBank mRNA | AF124251.1 | - | 7 | 1421 | ||
AJ420431.1 | - | 1 | 51 | |||
AK056068.1 | - | 6 | 1511 | |||
AK096983.1 | - | 4 | 804 | |||
AK128287.1 | - | 9 | 1913 | |||
AK291393.1 | - | 6 | 1475 | |||
AK292041.1 | - | 7 | 1437 | |||
AK292718.1 | - | 8 | 1853 | |||
AK294386.1 | - | 7 | 1374 | |||
AY358089.1 | - | 7 | 1491 | |||
AY358440.1 | - | 7 | 1410 | |||
BC027962.1 | - | 7 | 1380 | 5223012 | ||
BC032365.1 | - | 8 | 1857 | 4521962 | ||
BX647905.1 | - | 8 | 1806 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001252547, NM_013781, AB043953, AB030442, BC113203, and others.
Gene Ontology Categories:
