siRNA Details
Gene Symbol: SH2D3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142531.1 | NP_001136003.1 | 8 | 1575 | ||
NM_001142532.1 | NP_001136004.1 | 8 | 1824 | |||
NM_001142533.1 | NP_001136005.1 | 7 | 1712 | |||
NM_001142534.1 | NP_001136006.1 | 7 | 1684 | |||
NM_001252334.1 | NP_001239263.1 | 9 | 2081 | |||
NM_005489.3 | NP_005480.2 | 8 | 1710 | |||
NM_170600.2 | NP_733745.1 | 9 | 2183 | |||
XM_005251639.1 | XP_005251696.1 | 8 | 1691 | |||
XM_011518114.1 | XP_011516416.1 | 9 | 2167 | |||
XM_011518115.1 | XP_011516417.1 | 9 | 2127 | |||
XM_011518117.2 | XP_011516419.1 | 7 | 2803 | |||
XM_017014174.1 | XP_016869663.1 | 7 | 1964 | |||
GenBank mRNA | AF124251.1 | - | 8 | 1622 | ||
AJ420431.1 | - | 2 | 252 | |||
AK056068.1 | - | 7 | 1712 | |||
AK096983.1 | - | 5 | 1005 | |||
AK128287.1 | - | 10 | 2114 | |||
AK291393.1 | - | 7 | 1676 | |||
AK292041.1 | - | 8 | 1638 | |||
AK292718.1 | - | 9 | 2054 | |||
AK294386.1 | - | 8 | 1575 | |||
AY358089.1 | - | 8 | 1692 | |||
AY358440.1 | - | 8 | 1611 | |||
BC027962.1 | - | 8 | 1581 | 5223012 | ||
BC032365.1 | - | 9 | 2058 | 4521962 | ||
BX647905.1 | - | 9 | 2007 | |||
CU454732.1 | - | 1 | 59 | |||
CU455802.1 | - | 1 | 59 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
