siRNA Details
Gene Symbol: ODF2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007022.2 | NP_001007023.2 | 12 | 1556 | ||
NM_001184765.1 | NP_001171694.1 | 12 | 1563 | |||
NM_001184766.1 | NP_001171695.1 | 11 | 1476 | |||
NM_020729.2 | NP_065780.2 | 11 | 1476 | |||
XM_005271054.3 | XP_005271111.1 | 11 | 1474 | |||
XM_011541815.2 | XP_011540117.1 | 12 | 1561 | |||
XM_011541819.2 | XP_011540121.1 | 12 | 1561 | |||
XM_011541821.2 | XP_011540123.1 | 12 | 1561 | |||
XM_011541823.2 | XP_011540125.1 | 12 | 1561 | |||
XM_011541824.2 | XP_011540126.1 | 12 | 1561 | |||
XM_017001876.1 | XP_016857365.1 | 12 | 1353 | |||
XM_017001877.1 | XP_016857366.1 | 11 | 1266 | |||
XM_017001878.1 | XP_016857367.1 | 11 | 1444 | |||
XM_017001879.1 | XP_016857368.1 | 11 | 1474 | |||
XM_017001880.1 | XP_016857369.1 | 10 | 1357 | |||
XM_017001881.1 | XP_016857370.1 | 9 | 1229 | |||
XM_017001882.1 | XP_016857371.1 | 5 | 1674 | |||
GenBank mRNA | AB033055.1 | - | 12 | 1476 | ||
AK302792.1 | - | 11 | 1448 | |||
AK303209.1 | - | 11 | 1473 | |||
AK308351.1 | - | 8 | 1000 | |||
BC009779.1 | - | 7 | 874 | |||
BC029420.1 | - | 11 | 1194 | |||
BC091490.1 | - | 12 | 1462 | 30367572 | ||
BQ894001.1 | - | 223 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
