siRNA Details
Gene Symbol: DRICH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016449.3 | NP_057533.2 | 1 | 362 | ||
XM_011530206.1 | XP_011528508.1 | 1 | 362 | |||
XM_011530207.2 | XP_011528509.1 | 3 | 212 | |||
XM_011530208.2 | XP_011528510.1 | 1 | 362 | |||
XM_011530209.2 | XP_011528511.1 | 1 | 362 | |||
XM_011530210.2 | XP_011528512.1 | 2 | 43 | |||
XM_017028813.1 | XP_016884302.1 | 1 | 362 | |||
GenBank mRNA | AK310349.1 | - | 1 | 362 | ||
AL080197.1 | - | 1 | 342 | |||
BC056888.1 | - | 1 | 341 | 5165160 | ||
CR456370.1 | - | 1 | 89 | |||
DB052993.1 | - | 1 | 362 |
Gene Symbol: SRSF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK310317.1 | - | 362 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
