siRNA Details
Gene Symbol: UMODL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004416.2 | NP_001004416.2 | 23 | 4605 | ||
NM_001199527.1 | NP_001186456.1 | 22 | 5169 | |||
NM_001199528.2 | NP_001186457.2 | 23 | 4785 | |||
NM_173568.3 | NP_775839.3 | 22 | 4989 | |||
XM_011529797.1 | XP_011528099.1 | 20 | 4597 | |||
XM_017028506.1 | XP_016883995.1 | 22 | 4984 | |||
XM_017028507.1 | XP_016883996.1 | 22 | 7913 | |||
XM_017028508.1 | XP_016883997.1 | 12 | 2768 | |||
GenBank mRNA | AK093654.1 | - | 8 | 1847 | ||
AK124223.1 | - | 9 | 2349 | |||
AY771619.1 | - | 22 | 4989 | |||
AY771620.1 | - | 23 | 4605 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
