siRNA Details
Gene Symbol: UMODL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004416.2 | NP_001004416.2 | 23 | 4672 | ||
NM_001199527.1 | NP_001186456.1 | 22 | 5236 | |||
NM_001199528.2 | NP_001186457.2 | 23 | 4852 | |||
NM_173568.3 | NP_775839.3 | 22 | 5056 | |||
XM_011529797.1 | XP_011528099.1 | 20 | 4664 | |||
XM_017028506.1 | XP_016883995.1 | 22 | 5051 | |||
XM_017028507.1 | XP_016883996.1 | 22 | 7980 | |||
XM_017028508.1 | XP_016883997.1 | 12 | 2835 | |||
GenBank mRNA | AK093654.1 | - | 8 | 1914 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
