siRNA Details
Gene Symbol: EBF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005463.2 | NP_001005463.1 | 8 | 713 | ||
XM_005252667.3 | XP_005252724.1 | 8 | 1160 | |||
XM_005252668.3 | XP_005252725.1 | 8 | 1160 | |||
XM_005252669.3 | XP_005252726.1 | 8 | 1160 | |||
XM_006717739.3 | XP_006717802.1 | 8 | 1160 | |||
XM_006717740.3 | XP_006717803.1 | 8 | 1160 | |||
XM_006717741.3 | XP_006717804.1 | 8 | 1160 | |||
XM_006717742.3 | XP_006717805.1 | 8 | 1160 | |||
XM_006717743.3 | XP_006717806.1 | 8 | 1160 | |||
XM_006717744.3 | XP_006717807.1 | 8 | 1160 | |||
XM_011539574.2 | XP_011537876.1 | 8 | 1160 | |||
XM_011539575.2 | XP_011537877.1 | 3 | 154 | |||
XM_017016027.1 | XP_016871516.1 | 8 | 1160 | |||
GenBank mRNA | AL832828.1 | - | 8 | 640 | ||
BC126130.1 | - | 8 | 704 | 8991846 | ||
BC130479.1 | - | 8 | 704 | 40146510 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
