siRNA Details
Gene Symbol: ALDOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000034.3 | NP_000025.1 | 12 | 1748 | ||
NM_001127617.2 | NP_001121089.1 | 7 | 970 | |||
NM_001243177.1 | NP_001230106.1 | 8 | 1091 | |||
NM_184041.2 | NP_908930.1 | 7 | 937 | |||
NM_184043.2 | NP_908932.1 | 7 | 909 | |||
XM_011545768.2 | XP_011544070.1 | 9 | 1611 | |||
GenBank mRNA | AK026577.1 | - | 7 | 786 | ||
AK098778.1 | - | 10 | 1789 | |||
AK301993.1 | - | 8 | 903 | |||
BC000367.2 | - | 4 | 349 | |||
BC004333.1 | - | 12 | 1668 | 3630974 | ||
BC010660.1 | - | 7 | 855 | 3869603 | ||
BC012880.1 | - | 7 | 783 | 3459009 | ||
BC013614.1 | - | 7 | 866 | 3880857 | ||
BC015888.2 | - | 7 | 779 | 3911623 | ||
BC016170.1 | - | 4 | 349 | |||
BC016800.1 | - | 8 | 807 | 4072487 | ||
BU855133.1 | - | 2 | 56 | |||
BX647566.1 | - | 7 | 759 | |||
CR536528.1 | - | 6 | 661 | |||
CR541880.1 | - | 6 | 661 | |||
EF036496.1 | - | 4 | 309 | |||
EF036497.1 | - | 3 | 240 | |||
FJ474908.1 | - | 8 | 828 | |||
M11560.1 | - | 8 | 828 | |||
M21190.1 | - | 3 | 247 | |||
X05236.1 | - | 7 | 807 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
