siRNA Details
Gene Symbol: ALDOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000034.3 | NP_000025.1 | 14 | 2335 | ||
NM_001127617.2 | NP_001121089.1 | 9 | 1557 | |||
NM_001243177.1 | NP_001230106.1 | 10 | 1678 | |||
NM_184041.2 | NP_908930.1 | 9 | 1524 | |||
NM_184043.2 | NP_908932.1 | 9 | 1496 | |||
XM_011545768.2 | XP_011544070.1 | 11 | 2198 | |||
GenBank mRNA | AK026577.1 | - | 9 | 1373 | ||
AK098778.1 | - | 12 | 2376 | |||
AK301993.1 | - | 10 | 1490 | |||
BC000367.2 | - | 6 | 936 | |||
BC004333.1 | - | 14 | 2255 | 3630974 | ||
BC010660.1 | - | 9 | 1442 | 3869603 | ||
BC012880.1 | - | 9 | 1370 | 3459009 | ||
BC013614.1 | - | 9 | 1453 | 3880857 | ||
BC015888.2 | - | 9 | 1366 | 3911623 | ||
BC016170.1 | - | 6 | 936 | |||
BC016800.1 | - | 10 | 1394 | 4072487 | ||
BU855133.1 | - | 4 | 643 | |||
BX647566.1 | - | 9 | 1346 | |||
FJ474908.1 | - | 10 | 1416 | |||
M11560.1 | - | 10 | 1416 | |||
M21190.1 | - | 5 | 834 | |||
X05236.1 | - | 9 | 1392 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
