siRNA Details
Gene Symbol: ALDOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000034.3 | NP_000025.1 | 7 | 1137 | ||
NM_001127617.2 | NP_001121089.1 | 2 | 359 | |||
NM_001243177.1 | NP_001230106.1 | 3 | 480 | |||
NM_184041.2 | NP_908930.1 | 2 | 326 | |||
NM_184043.2 | NP_908932.1 | 2 | 298 | |||
XM_011545768.2 | XP_011544070.1 | 4 | 1000 | |||
GenBank mRNA | AK026577.1 | - | 2 | 176 | ||
AK098778.1 | - | 5 | 1178 | |||
AK301993.1 | - | 3 | 292 | |||
BC004333.1 | - | 7 | 1057 | 3630974 | ||
BC010660.1 | - | 2 | 244 | 3869603 | ||
BC012880.1 | - | 2 | 172 | 3459009 | ||
BC013614.1 | - | 2 | 255 | 3880857 | ||
BC015888.2 | - | 2 | 168 | 3911623 | ||
BC016800.1 | - | 3 | 196 | 4072487 | ||
BE893518.1 | - | 2 | 302 | |||
BQ719865.1 | - | 2 | 287 | |||
BX647566.1 | - | 2 | 148 | |||
CB151262.1 | - | 2 | 297 | |||
CR536528.1 | - | 1 | 50 | |||
CR541880.1 | - | 1 | 50 | |||
DA138954.1 | - | 2 | 326 | |||
DA893541.1 | - | 2 | 206 | |||
DA899482.1 | - | 2 | 359 | |||
FJ474908.1 | - | 3 | 217 | |||
M11560.1 | - | 3 | 217 | |||
X05236.1 | - | 2 | 196 |
Gene Symbol: ALDOAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | M21191.1 | - | LIT2112 ALDOAP2 |
1 | 300 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001177308, NM_001177307, NM_001277340, NM_007438, AK006425, and others.
Gene Ontology Categories:
