siRNA Details
Gene Symbol: CRTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098482.1 | NP_001091952.1 | 5 | 543 | ||
NM_015321.2 | NP_056136.2 | 4 | 495 | |||
XM_005259833.3 | XP_005259890.1 | 5 | 504 | |||
XM_005259834.1 | XP_005259891.1 | 4 | 489 | |||
XM_005259835.3 | XP_005259892.1 | 5 | 503 | |||
XM_005259836.3 | XP_005259893.1 | 5 | 503 | |||
XM_006722710.3 | XP_006722773.1 | 5 | 503 | |||
XM_011527842.2 | XP_011526144.1 | 5 | 504 | |||
XM_017026536.1 | XP_016882025.1 | 5 | 504 | |||
XM_017026537.1 | XP_016882026.1 | 5 | 504 | |||
XM_017026538.1 | XP_016882027.1 | 5 | 504 | |||
GenBank mRNA | AB014516.1 | - | 5 | 408 | ||
AK024089.1 | - | 4 | 495 | |||
AU118537.1 | - | 4 | 495 | |||
AY040323.1 | - | 4 | 495 | |||
AY360171.1 | - | 5 | 480 | |||
BC023614.2 | - | 4 | 318 | |||
BC028050.1 | - | 4 | 427 | 4938995 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
