siRNA Details
Gene Symbol: CRTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098482.1 | NP_001091952.1 | 15 | 1859 | ||
NM_015321.2 | NP_056136.2 | 14 | 1811 | |||
XM_005259833.3 | XP_005259890.1 | 14 | 1733 | |||
XM_005259834.1 | XP_005259891.1 | 13 | 1718 | |||
XM_005259835.3 | XP_005259892.1 | 15 | 1561 | |||
XM_005259836.3 | XP_005259893.1 | 14 | 1456 | |||
XM_006722710.3 | XP_006722773.1 | 13 | 1405 | |||
XM_011527842.2 | XP_011526144.1 | 14 | 1715 | |||
GenBank mRNA | AB014516.1 | - | 15 | 1724 | ||
AK024089.1 | - | 13 | 1724 | |||
AY040323.1 | - | 13 | 1724 | |||
AY360171.1 | - | 15 | 1796 | |||
BC017075.2 | - | 10 | 1292 | |||
BC023614.2 | - | 14 | 1634 | |||
BC028050.1 | - | 14 | 1743 | 4938995 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
