siRNA Details
Gene Symbol: SMIM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032947.4 | NP_116565.3 | 2 | 2312 | ||
GenBank mRNA | AF313413.1 | - | 2 | 1434 | ||
AK027847.1 | - | 2 | 1499 | |||
AK125286.1 | - | 2 | 2312 | |||
BC009719.1 | - | 2 | 1428 | |||
BC106040.1 | - | 2 | 1965 | 4940258 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
