siRNA Details
Gene Symbol: CEP192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032142.3 | NP_115518.3 | 25 | 4970 | ||
XM_005258107.3 | XP_005258164.1 | 25 | 5025 | |||
XM_005258109.3 | XP_005258166.1 | 24 | 4815 | |||
XM_005258110.1 | XP_005258167.1 | 12 | 3209 | |||
XM_006722326.3 | XP_006722389.1 | 25 | 5025 | |||
XM_006722327.3 | XP_006722390.1 | 24 | 4815 | |||
XM_006722330.3 | XP_006722393.1 | 19 | 4008 | |||
XM_011525673.2 | XP_011523975.1 | 25 | 5025 | |||
XM_011525675.2 | XP_011523977.1 | 22 | 4574 | |||
XM_017025803.1 | XP_016881292.1 | 24 | 4860 | |||
XM_017025804.1 | XP_016881293.1 | 19 | 4008 | |||
XM_017025805.1 | XP_016881294.1 | 14 | 3349 | |||
XM_017025806.1 | XP_016881295.1 | 22 | 4455 | |||
XM_017025807.1 | XP_016881296.1 | 18 | 4067 | |||
GenBank mRNA | AB046789.1 | - | 6 | 2149 | ||
AF318363.1 | - | 5 | 534 | |||
AK074074.1 | - | 10 | 2474 | |||
AL136818.1 | - | 10 | 2623 | |||
AL833494.2 | - | 16 | 3688 | |||
BC144480.1 | - | 15 | 3506 | 9053007 | ||
BC144481.1 | - | 16 | 3716 | 9053008 | ||
BX648232.1 | - | 19 | 5629 | |||
BX649151.1 | - | 12 | 3209 | |||
KJ567064.1 | - | 24 | 4890 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
