siRNA Details
Gene Symbol: ZSCAN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007072.1 | NP_001007073.1 | 2 | 298 | ||
NM_017894.5 | NP_060364.4 | 2 | 298 | |||
NM_181877.3 | NP_870992.2 | 2 | 298 | |||
XM_017022392.1 | XP_016877881.1 | 2 | 330 | |||
XM_017022393.1 | XP_016877882.1 | 2 | 330 | |||
GenBank mRNA | AK000602.1 | - | 2 | 271 | ||
AK128624.1 | - | 2 | 364 | |||
AK291224.1 | - | 2 | 296 | |||
AY262259.1 | - | 2 | 266 | |||
AY279349.1 | - | 2 | 266 | |||
BC041620.1 | - | 2 | 298 | |||
BC136342.1 | - | 1 | 99 | 9020329 | ||
BC143992.1 | - | 1 | 99 | 9052510 | ||
BC143993.1 | - | 1 | 99 | 9052511 | ||
BC143994.1 | - | 1 | 99 | 9052512 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
