siRNA Details
Gene Symbol: AXDND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144696.5 | NP_653297.3 | 19 | 2657 | ||
NR_073544.1 | 18 | 2296 | ||||
XM_011509165.1 | XP_011507467.1 | 19 | 2636 | |||
XM_011509166.2 | XP_011507468.1 | 19 | 2683 | |||
XM_011509167.2 | XP_011507469.1 | 19 | 2798 | |||
XM_011509168.1 | XP_011507470.1 | 19 | 2640 | |||
XM_011509169.1 | XP_011507471.1 | 18 | 2429 | |||
XM_011509170.1 | XP_011507472.1 | 19 | 2636 | |||
XM_011509171.1 | XP_011507473.1 | 18 | 2454 | |||
XM_011509174.1 | XP_011507476.1 | 19 | 2636 | |||
XM_011509175.1 | XP_011507477.1 | 19 | 2636 | |||
XM_011509178.2 | XP_011507480.1 | 19 | 2636 | |||
XM_011509179.2 | XP_011507481.1 | 14 | 1744 | |||
XM_011509180.1 | XP_011507482.1 | 19 | 2636 | |||
XM_011509181.2 | XP_011507483.1 | 9 | 1253 | |||
XM_017000257.1 | XP_016855746.1 | 13 | 1604 | |||
XM_017000258.1 | XP_016855747.1 | 13 | 1636 | |||
XM_017000259.1 | XP_016855748.1 | 5 | 806 | |||
GenBank mRNA | AK058167.1 | - | 1 | 14 | ||
AL833914.1 | - | 17 | 2084 | |||
BC146823.1 | - | 18 | 2297 | 8860438 | ||
BX647935.1 | - | 19 | 2659 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
