siRNA Details
Gene Symbol: COL4A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000092.4 | NP_000083.3 | 47 | 5258 | ||
XM_005246281.3 | XP_005246338.1 | 47 | 4792 | |||
XM_005246282.3 | XP_005246339.1 | 41 | 4375 | |||
XM_006712246.3 | XP_006712309.1 | 46 | 4741 | |||
XM_011510557.2 | XP_011508859.1 | 48 | 4849 | |||
XM_011510558.2 | XP_011508860.1 | 46 | 4822 | |||
XM_011510559.2 | XP_011508861.1 | 47 | 4930 | |||
XM_011510560.2 | XP_011508862.1 | 47 | 4930 | |||
XM_011510561.2 | XP_011508863.1 | 47 | 4930 | |||
XM_011510562.2 | XP_011508864.1 | 47 | 4930 | |||
XM_011510572.2 | XP_011508874.1 | 26 | 3155 | |||
XM_017003296.1 | XP_016858785.1 | 47 | 4930 | |||
XM_017003297.1 | XP_016858786.1 | 47 | 4813 | |||
XM_017003298.1 | XP_016858787.1 | 47 | 4930 | |||
GenBank mRNA | D17391.1 | - | 9 | 951 | ||
X81053.1 | - | 47 | 4812 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
