siRNA Details
Gene Symbol: SNX20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144972.1 | NP_001138444.1 | 3 | 396 | ||
NM_153337.2 | NP_699168.1 | 3 | 396 | |||
NM_182854.3 | NP_878274.1 | 3 | 396 | |||
GenBank mRNA | AK055837.1 | - | 3 | 361 | ||
AK292650.1 | - | 3 | 355 | |||
AK292705.1 | - | 3 | 361 | |||
AY302441.1 | - | 3 | 298 | |||
BC027944.1 | - | 3 | 345 | |||
BC063423.1 | - | 3 | 337 | |||
BC119626.1 | - | 3 | 249 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
