siRNA Details
Gene Symbol: CEACAM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127893.1 | NP_001121365.1 | 8 | 2006 | ||
NM_020219.3 | NP_064604.2 | 8 | 2009 | |||
GenBank mRNA | AK027157.1 | - | 8 | 1723 | ||
AK128234.1 | - | 4 | 3033 | |||
AL390165.1 | - | 1 | 392 | |||
AY358394.1 | - | 8 | 1739 | |||
BC083499.1 | - | 8 | 2006 | 30390224 | ||
KJ851726.1 | - | 8 | 1554 | |||
KJ851727.1 | - | 8 | 1557 | |||
KJ851728.1 | - | 9 | 1589 | |||
KJ851729.1 | - | 9 | 1592 | |||
KJ851730.1 | - | 9 | 1593 | |||
KJ851731.1 | - | 9 | 1596 | |||
KJ851732.1 | - | 7 | 1442 | |||
KJ851733.1 | - | 7 | 1445 | |||
KJ851734.1 | - | 7 | 1470 | |||
KJ851735.1 | - | 7 | 1473 | |||
KJ851736.1 | - | 8 | 1505 | |||
KJ851737.1 | - | 8 | 1508 | |||
KJ851738.1 | - | 8 | 1509 | |||
KJ851739.1 | - | 8 | 1512 | |||
KJ851740.1 | - | 5 | 1309 | |||
KJ851741.1 | - | 5 | 1312 | |||
KJ851742.1 | - | 7 | 1375 | |||
KJ851743.1 | - | 7 | 1378 | |||
KJ851744.1 | - | 7 | 1403 | |||
KJ851745.1 | - | 7 | 1406 | |||
KJ851746.1 | - | 8 | 1438 | |||
KJ851747.1 | - | 8 | 1441 | |||
KJ851748.1 | - | 8 | 1442 | |||
KJ851749.1 | - | 8 | 1445 | |||
KJ851750.1 | - | 6 | 1291 | |||
KJ851751.1 | - | 6 | 1294 | |||
KJ851752.1 | - | 6 | 1319 | |||
KJ851753.1 | - | 6 | 1322 | |||
KJ851754.1 | - | 7 | 1354 | |||
KJ851755.1 | - | 7 | 1357 | |||
KJ851756.1 | - | 7 | 1358 | |||
KJ851757.1 | - | 7 | 1361 | |||
KJ851758.1 | - | 4 | 1158 | |||
KJ851759.1 | - | 4 | 1161 | |||
KJ851760.1 | - | 9 | 1660 | |||
KJ851761.1 | - | 9 | 1663 | |||
KJ851762.1 | - | 9 | 1688 | |||
KJ851763.1 | - | 9 | 1691 | |||
KJ851764.1 | - | 10 | 1723 | |||
KJ851765.1 | - | 10 | 1726 | |||
KJ851766.1 | - | 11 | 1727 | |||
KJ851767.1 | - | 11 | 1730 | |||
KJ851768.1 | - | 8 | 1576 | |||
KJ851769.1 | - | 8 | 1579 | |||
KJ851770.1 | - | 8 | 1604 | |||
KJ851771.1 | - | 8 | 1607 | |||
KJ851772.1 | - | 9 | 1639 | |||
KJ851773.1 | - | 9 | 1642 | |||
KJ851774.1 | - | 9 | 1643 | |||
KJ851775.1 | - | 9 | 1646 | |||
KJ851776.1 | - | 6 | 1443 | |||
KJ851777.1 | - | 6 | 1446 | |||
KJ851778.1 | - | 8 | 1509 | |||
KJ851779.1 | - | 8 | 1512 | |||
KJ851780.1 | - | 8 | 1537 | |||
KJ851781.1 | - | 8 | 1540 | |||
KJ851782.1 | - | 9 | 1572 | |||
KJ851783.1 | - | 9 | 1575 | |||
KJ851784.1 | - | 9 | 1576 | |||
KJ851785.1 | - | 9 | 1579 | |||
KJ851786.1 | - | 7 | 1425 | |||
KJ851787.1 | - | 7 | 1428 | |||
KJ851788.1 | - | 7 | 1453 | |||
KJ851789.1 | - | 7 | 1456 | |||
KJ851790.1 | - | 8 | 1488 | |||
KJ851791.1 | - | 8 | 1491 | |||
KJ851792.1 | - | 8 | 1492 | |||
KJ851793.1 | - | 8 | 1495 | |||
KJ851794.1 | - | 5 | 1292 | |||
KJ851795.1 | - | 5 | 1295 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
