siRNA Details
Gene Symbol: CEACAM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127893.1 | NP_001121365.1 | 8 | 2099 | ||
NM_020219.3 | NP_064604.2 | 8 | 2102 | |||
GenBank mRNA | AK027157.1 | - | 8 | 1816 | ||
AK128234.1 | - | 4 | 3126 | |||
AL390165.1 | - | 1 | 485 | |||
AY358394.1 | - | 8 | 1832 | |||
BC083499.1 | - | 8 | 2099 | 30390224 | ||
KJ851726.1 | - | 8 | 1647 | |||
KJ851727.1 | - | 8 | 1650 | |||
KJ851728.1 | - | 9 | 1682 | |||
KJ851729.1 | - | 9 | 1685 | |||
KJ851730.1 | - | 9 | 1686 | |||
KJ851731.1 | - | 9 | 1689 | |||
KJ851732.1 | - | 7 | 1535 | |||
KJ851733.1 | - | 7 | 1538 | |||
KJ851734.1 | - | 7 | 1563 | |||
KJ851735.1 | - | 7 | 1566 | |||
KJ851736.1 | - | 8 | 1598 | |||
KJ851737.1 | - | 8 | 1601 | |||
KJ851738.1 | - | 8 | 1602 | |||
KJ851739.1 | - | 8 | 1605 | |||
KJ851740.1 | - | 5 | 1402 | |||
KJ851741.1 | - | 5 | 1405 | |||
KJ851742.1 | - | 7 | 1468 | |||
KJ851743.1 | - | 7 | 1471 | |||
KJ851744.1 | - | 7 | 1496 | |||
KJ851745.1 | - | 7 | 1499 | |||
KJ851746.1 | - | 8 | 1531 | |||
KJ851747.1 | - | 8 | 1534 | |||
KJ851748.1 | - | 8 | 1535 | |||
KJ851749.1 | - | 8 | 1538 | |||
KJ851750.1 | - | 6 | 1384 | |||
KJ851751.1 | - | 6 | 1387 | |||
KJ851752.1 | - | 6 | 1412 | |||
KJ851753.1 | - | 6 | 1415 | |||
KJ851754.1 | - | 7 | 1447 | |||
KJ851755.1 | - | 7 | 1450 | |||
KJ851756.1 | - | 7 | 1451 | |||
KJ851757.1 | - | 7 | 1454 | |||
KJ851758.1 | - | 4 | 1251 | |||
KJ851759.1 | - | 4 | 1254 | |||
KJ851760.1 | - | 9 | 1753 | |||
KJ851761.1 | - | 9 | 1756 | |||
KJ851762.1 | - | 9 | 1781 | |||
KJ851763.1 | - | 9 | 1784 | |||
KJ851764.1 | - | 10 | 1816 | |||
KJ851765.1 | - | 10 | 1819 | |||
KJ851766.1 | - | 11 | 1820 | |||
KJ851767.1 | - | 11 | 1823 | |||
KJ851768.1 | - | 8 | 1669 | |||
KJ851769.1 | - | 8 | 1672 | |||
KJ851770.1 | - | 8 | 1697 | |||
KJ851771.1 | - | 8 | 1700 | |||
KJ851772.1 | - | 9 | 1732 | |||
KJ851773.1 | - | 9 | 1735 | |||
KJ851774.1 | - | 9 | 1736 | |||
KJ851775.1 | - | 9 | 1739 | |||
KJ851776.1 | - | 6 | 1536 | |||
KJ851777.1 | - | 6 | 1539 | |||
KJ851778.1 | - | 8 | 1602 | |||
KJ851779.1 | - | 8 | 1605 | |||
KJ851780.1 | - | 8 | 1630 | |||
KJ851781.1 | - | 8 | 1633 | |||
KJ851782.1 | - | 9 | 1665 | |||
KJ851783.1 | - | 9 | 1668 | |||
KJ851784.1 | - | 9 | 1669 | |||
KJ851785.1 | - | 9 | 1672 | |||
KJ851786.1 | - | 7 | 1518 | |||
KJ851787.1 | - | 7 | 1521 | |||
KJ851788.1 | - | 7 | 1546 | |||
KJ851789.1 | - | 7 | 1549 | |||
KJ851790.1 | - | 8 | 1581 | |||
KJ851791.1 | - | 8 | 1584 | |||
KJ851792.1 | - | 8 | 1585 | |||
KJ851793.1 | - | 8 | 1588 | |||
KJ851794.1 | - | 5 | 1385 | |||
KJ851795.1 | - | 5 | 1388 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
