siRNA Details
Gene Symbol: GNG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243773.1 | NP_001230702.1 | 3 | 627 | ||
NM_001243774.1 | NP_001230703.1 | 2 | 590 | |||
NM_053064.4 | NP_444292.1 | 3 | 631 | |||
XM_006720173.2 | XP_006720236.1 | 3 | 392 | |||
XM_011536846.2 | XP_011535148.1 | 3 | 169 | |||
XM_017021376.1 | XP_016876865.1 | 4 | 413 | |||
XM_017021377.1 | XP_016876866.1 | 5 | 870 | |||
GenBank mRNA | AF493870.1 | - | 1 | 74 | ||
AK001024.1 | - | 3 | 289 | |||
AK092371.1 | - | 2 | 480 | |||
AL832878.1 | - | 3 | 266 | |||
BC020774.1 | - | 3 | 203 | 4243482 | ||
BC041446.1 | - | 5 | 801 | |||
BC060856.1 | - | 2 | 169 | 5276390 | ||
BE245271.1 | - | 2 | 283 | |||
DA226565.1 | - | 3 | 197 | |||
DA785895.1 | - | 3 | 196 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
