siRNA Details
Gene Symbol: WDR25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161476.1 | NP_001154948.1 | 2 | 560 | ||
NM_024515.4 | NP_078791.3 | 2 | 691 | |||
XM_005268056.4 | XP_005268113.1 | 2 | 567 | |||
XM_011537150.2 | XP_011535452.1 | 2 | 814 | |||
GenBank mRNA | AK291960.1 | - | 2 | 523 | ||
AY295082.1 | - | 2 | 574 | |||
BC007953.2 | - | 1 | 492 | |||
BC067872.1 | - | 2 | 646 | |||
BX248767.1 | - | 2 | 553 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
