siRNA Details
Gene Symbol: GIMAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244071.1 | NP_001231000.1 | 3 | 2943 | ||
NM_001244072.1 | NP_001231001.1 | 3 | 3377 | |||
NM_024711.5 | NP_078987.3 | 3 | 3167 | |||
GenBank mRNA | AK026343.1 | - | 4 | 1648 | ||
AK026396.1 | - | 1 | 1700 | |||
AK074044.1 | - | 2 | 4359 | |||
BC060760.1 | - | 3 | 2829 | 30336063 | ||
CR749355.1 | - | 3 | 2884 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
