siRNA Details
Gene Symbol: GIMAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244071.1 | NP_001231000.1 | 3 | 786 | ||
NM_001244072.1 | NP_001231001.1 | 3 | 1220 | |||
NM_024711.5 | NP_078987.3 | 3 | 1010 | |||
GenBank mRNA | AJ580952.1 | - | 3 | 484 | ||
AK026343.1 | - | 3 | 457 | |||
AK074044.1 | - | 2 | 2201 | |||
AK294989.1 | - | 3 | 855 | |||
AK300445.1 | - | 3 | 726 | |||
AK314667.1 | - | 3 | 714 | |||
AL541003.3 | - | 3 | 717 | |||
BC060760.1 | - | 3 | 671 | 30336063 | ||
BC074744.2 | - | 3 | 560 | 30915204 | ||
CR749355.1 | - | 3 | 747 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
