siRNA Details
Gene Symbol: KLHL21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324309.1 | NP_001311238.1 | 2 | 1494 | ||
NM_014851.3 | NP_055666.2 | 2 | 1494 | |||
GenBank mRNA | AB007938.1 | - | 2 | 1597 | ||
AK075305.1 | - | 2 | 1465 | |||
AK090472.1 | - | 2 | 1263 | |||
AK307564.1 | - | 2 | 454 | |||
BC034039.1 | - | 2 | 1465 | 4811534 | ||
BC064980.1 | - | 2 | 1447 | |||
BC091648.1 | - | 2 | 1447 | 6373903 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
